Hydroxypropyl cellulose, 3-6 mPa.s (2% in H₂O 20 °C)
Supplier: TCI America
H0473-25G
H0473-500G
TCH0473-025GEA
56.28
CAD
TCH0473-025G
TCH0473-500G
Hydroxypropyl cellulose, 3-6 mPa.s (2% in H₂O 20 °C)
Hydroxypropyl cellulose
CAS Number: 9004-64-2
MDL Number: MFCD00132688
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)
MDL Number:
MFCD00132688 CAS Number: 9004-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...