1-(Heptafluorobutyryl)imidazole ≥97.0%

Supplier: TCI America

Synonyms: 1-(Perfluorobutanoyl)-1H-imidazole, N-Heptafluorobutyrylimidazole

H0467-5G H0467-25G
TCH0467-005GEA 175.38 CAD
TCH0467-005G TCH0467-25G
1-(Heptafluorobutyryl)imidazole ≥97.0%
1-(Heptafluorobutyryl)imidazole

[Acylating Agent]
CAS Number: 32477-35-3
MDL Number: MFCD00014503
Molecular Formula: C7H3F7N2O
Molecular Weight: 264.10
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 161
Melting point (°C): 13
Flash Point (°C): 77
Specific Gravity (20/20): 1.58
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Other Fluorinated Group Introducing Agents, Analytical Science, Acylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: 1-(Perfluorobutyryl)imidazole

Formula: C₇H₃F₇N₂O
MW: 264.10 g/mol
Boiling Pt: 161 °C
Melting Pt: 13 °C
Density: 1.58 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00014503
CAS Number: 32477-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ed3a1870d875fece25cf7f52104650269347d27&vid=3fb7b172aefd86b3a205c2a5baeaee27b306c080&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR