2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate ≥98.0% (by titrimetric analysis) (contains 5-10% Isopropyl Alcohol)

Supplier: TCI America

H0466-25G H0466-500G
TCH0466-025GEA 79 CAD
TCH0466-025G TCH0466-500G
2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate ≥98.0% (by titrimetric analysis) (contains 5-10% Isopropyl Alcohol)
2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate

CAS Number: 4065-45-6
MDL Number: MFCD00024962
Molecular Formula: C14H12O6S
Molecular Weight: 308.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic Acid, Sulisobenzone

Formula: C₁₄H₁₂O₆S·H₂O
MDL Number: MFCD00024962
CAS Number: 4065-45-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b6eed99d4e189eec26d7710756354b20bcd050d&vid=1268a4ab1d8c5b0c861369cb85854282b0fec79a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR