1-Hydroxyethylferrocene ≥95.0% (by titrimetric analysis)

Supplier: TCI America

H0463-5G H0463-1G
TCH0463-005GEA 208.56 CAD
TCH0463-005G TCH0463-001G
1-Hydroxyethylferrocene ≥95.0% (by titrimetric analysis)
1-Hydroxyethylferrocene

CAS Number: 1277-49-2
MDL Number: MFCD00013752
Molecular Formula: C12H14FeO
Molecular Weight: 230.09
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: 1-Ferrocenylethanol

Formula: C₁₂H₅FeO
Melting Pt: 78 °C
MDL Number: MFCD00013752
CAS Number: 1277-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df1fc4f25c34e043319f50d032b0246badd28ddc&vid=df1fc4f25c34e043319f50d032b0246badd28ddc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR