Sodium 1-hexadecanesulfonate

Supplier: TCI America

Synonyms: Hexadecane-1-sulfonic acid sodium salt, Sodium hexadecane-1-sulphonate, Hexadecane-1-sulphonic acid sodium salt, Sodium hexadecane-1-sulfonate

H0415-10G H0415-25G
TCH0415-010GEA 99.54 CAD
TCH0415-010G TCH0415-25G
Sodium 1-hexadecanesulfonate
Sodium 1-hexadecanesulphonate

CAS Number: 15015-81-3
MDL Number: MFCD00007528
Molecular Formula: C16H34O3S
Molecular Weight: 328.49
Form: Crystal
Color: White


Other Product Names / Synonyms: Cetylsulfonic Acid Sodium Salt, Hexadecylsulfonic Acid Sodium Salt, Sodium Cetylsulfonate, 1-Hexadecanesulfonic Acid Sodium Salt

Formula: C₁₆H₃₃NaSO₃
MDL Number: MFCD00007528
CAS Number: 15015-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63cc5535bb7b50e1f0f3fc443f57cfc168a7ecb9&vid=63cc5535bb7b50e1f0f3fc443f57cfc168a7ecb9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR