Het anhydride ≥95.0%

Supplier: TCI America

Synonyms: Chlorendic Anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic Anhydride

H0349-25G H0349-500G
TCH0349-025GEA 68.71 CAD
TCH0349-025G TCH0349-500G
Het anhydride ≥95.0%
Het anhydride

CAS Number: 115-27-5
MDL Number: MFCD00080438
Molecular Formula: C9H2Cl6O3
Molecular Weight: 370.81
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 238
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chlorendic Anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic Anhydride

Formula: C₉H₂Cl₆O₃
MW: 370.81 g/mol
Melting Pt: 238 °C
MDL Number: MFCD00080438
CAS Number: 115-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05ffa93de42c13f4d1b831404e212b1c91278e21&vid=05ffa93de42c13f4d1b831404e212b1c91278e21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR