12-Hydroxystearic acid ≥80.0%

Supplier: TCI America

Synonyms: 12-Hydroxyoctadecanoic Acid

H0308-25G H0308-500G
TCH0308-25GEA 53.72 CAD
TCH0308-25G TCH0308-500G
12-Hydroxystearic acid ≥80.0%
12-Hydroxystearic acid

CAS Number: 106-14-9
MDL Number: MFCD00004592
Molecular Formula: C18H36O3
Molecular Weight: 300.48
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 78
Flash Point (°C): 200


Other Product Names / Synonyms: 12-Hydroxyoctadecanoic Acid

Formula: C₁₈H₃₆O₃
Boiling Pt: 180 °C
Melting Pt: 78 °C
Flash Pt: 200 °C
MDL Number: MFCD00004592
CAS Number: 106-14-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7744feaaf0029b456dfefdc79e2c323c3b6d6eff&vid=7744feaaf0029b456dfefdc79e2c323c3b6d6eff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR