2-Mercaptopyridine-N-oxide ≥95.0%

Supplier: TCI America

Synonyms: 2-Mercaptopyridine 1-oxide

H0302-5G H0302-25G
TCH0302-005GEA 84.37 CAD
TCH0302-005G TCH0302-025G
2-Mercaptopyridine-N-oxide ≥95.0%
2-Mercaptopyridine N-oxide

CAS Number: 1121-31-9
MDL Number: MFCD00006196
Molecular Formula: C5H5NOS
Molecular Weight: 127.16
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 72
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Hydroxy-2-pyridinethione

Formula: C₅H₅NOS
Melting Pt: 72 °C
MDL Number: MFCD00006196
CAS Number: 1121-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1f23c18830ad563ac8a1bc20ca0c47c3582e6e7&vid=c1f23c18830ad563ac8a1bc20ca0c47c3582e6e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR