2-Hydroxy-4-octyloxybenzophenone ≥98.0%

Supplier: TCI America

H0288-25G H0288-500G
TCH0288-025GEA 36.96 CAD
TCH0288-025G TCH0288-500G
2-Hydroxy-4-octyloxybenzophenone ≥98.0%
2-Hydroxy-4-octyloxybenzophenone

CAS Number: 1843-05-6
MDL Number: MFCD00027327
Molecular Formula: C21H26O3
Molecular Weight: 326.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 48

Formula: C₂₁H₂₆O₃
Melting Pt: 48 °C
MDL Number: MFCD00027327
CAS Number: 1843-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cb6ae93748cd1b6b0050513623931755a5f52ea&vid=7cb6ae93748cd1b6b0050513623931755a5f52ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR