7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

Supplier: TCI America

Synonyms: 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 4-Hydroxy-6-methyl-1,3,3a,7-tetraazaindene

H0273-25G H0273-500G
TCH0273-025GEA 79 CAD
TCH0273-025G TCH0273-500G
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

CAS Number: 2503-56-2
MDL Number: MFCD00005555
Molecular Formula: C6H6N4O
Molecular Weight: 150.14
Form: Crystal
Color: White
Melting point (°C): 226


Other Product Names / Synonyms: 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, 4-Hydroxy-6-methyl-1,3,3a,7-tetraazaindene

Formula: C₆H₆N₄O
MW: 150.14 g/mol
Melting Pt: 226 °C
MDL Number: MFCD00005555
CAS Number: 2503-56-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f4f7b72477f85c3de217e63923c17fd94aea955&vid=0f4f7b72477f85c3de217e63923c17fd94aea955&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR