2-(Hydroxymethyl)-2-nitro-1,3-propanediol ≥99.0%

Supplier: TCI America

Synonyms: 2-(Hydroxymethyl)-2-nitropropane-1,3-diol, Tris(hydroxymethyl)nitromethane, Trimethylolnitromethane

H0271-500G H0271-25G
TCH0271-500GEA 259.12 CAD
TCH0271-500G TCH0271-25G
2-(Hydroxymethyl)-2-nitro-1,3-propanediol ≥99.0%
2-(Hydroxymethyl)-2-nitro-1,3-propanediol

CAS Number: 126-11-4
MDL Number: MFCD00007395
Molecular Formula: C4H9NO5
Molecular Weight: 151.12
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 214


Other Product Names / Synonyms: Trimethylolnitromethane, Tris(hydroxymethyl)nitromethane

Formula: C₄H₉NO₅
MW: 151.12 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00007395
CAS Number: 126-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3617b67e856b49b59a02ad4c6ddf2179f05de6c6&vid=3617b67e856b49b59a02ad4c6ddf2179f05de6c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR