N,N'-Bis(salicylidene)ethylenediamine ≥99.0%

Supplier: TCI America

Synonyms: Ethylenebis(salicylimine), 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))diphenol, SALEN

H0199-500G H0199-25G
TCH0199-500GEA 570.38 CAD
TCH0199-500G TCH0199-025G
N,N'-Bis(salicylidene)ethylenediamine ≥99.0%
N,N'-Bis(salicylidene)ethylenediamine

CAS Number: 94-93-9
MDL Number: MFCD00002244
Molecular Formula: C16H16N2O2
Molecular Weight: 268.32
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 127


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: N,N'-Bis(2-hydroxybenzylidene)ethylenediamine, N,N'-Disalicylalethylenediamine, Salen

Formula: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
Melting Pt: 127 °C
MDL Number: MFCD00002244
CAS Number: 94-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b3a7c50d53239a54d022881af2edcc1a745ebf4&vid=9b3a7c50d53239a54d022881af2edcc1a745ebf4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR