DL-Homocysteic acid ≥97.0%

Supplier: TCI America

Synonyms: 2-Amino-4-sulfobutanoic acid, 2-Amino-4-sulphobutanoic acid

H0158-5G H0158-1G
TCH0158-5GEA 556.16 CAD
TCH0158-5G TCH0158-001G
DL-Homocysteic acid ≥97.0%
DL-Homocysteic acid

CAS Number: 504-33-6
MDL Number: MFCD00007536
Molecular Formula: C4H9NO5S
Molecular Weight: 183.18
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Formula: C₄H₉NO₅S
MDL Number: MFCD00007536
CAS Number: 504-33-6

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR