2,5-Hexanediol ≥98.0%
Supplier: TCI America
H0100-25ML
H0100-5ML
TCH0100-025MLEA
164.41
CAD
TCH0100-025ML
TCH0100-5ML
2,5-Hexanediol ≥98.0%
2,5-Hexanediol
CAS Number: 2935-44-6
MDL Number: MFCD00004557
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Flash Point (°C): 110
Specific Gravity (20/20): 0.96
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,5-Dihydroxyhexane
Formula:
C₆H₁₄O₂ Boiling Pt: 221 °C Density: 0.96 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004557 CAS Number: 2935-44-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...