Guanidinium thiocyanate ≥98.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

G0360-25G G0360-5G
TCG0360-25GEA 48.98 CAD
TCG0360-25G TCG0360-5G
Guanidinium thiocyanate ≥98.0% (by titrimetric analysis) for biochemical research
Guanidinium thiocyanate

[for Biochemical Research]
CAS Number: 593-84-0
MDL Number: MFCD00013027
Molecular Formula: CH5N3
Molecular Weight: 118.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 120

Formula: CH₅N₃·CHNS
Melting Pt: 120 °C
MDL Number: MFCD00013027
CAS Number: 593-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4054cd96ff295b715f4d11a8f5eabcac525e2c37&vid=91c22e9f84ce4c6e29fef9c28580cdc1961e21a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR