Glycerine ≥99.0%

Supplier: TCI America

Synonyms: Propane-1,2,3-triolGlycerol1,2,3-Propanetriol

G0316-1G
TCG0316-1GEA 67.07 CAD
TCG0316-1G
Glycerine ≥99.0%
Glycerine

CAS Number: 56-81-5
MDL Number: MFCD00004722
Molecular Formula: C3H8O3
Molecular Weight: 92.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 290
Flash Point (°C): 177
Specific Gravity (20/20): 1.26


Product Categories: Analytical Science, Reagents for Gel & Buffer Preparation, etc. (Reagents for Electrophoresis)
Other Product Names / Synonyms: Glycerin

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C
Density: 1.26 (20 °C)
Flash Pt: 177 °C
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://ca.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR