D-Glucoronic acid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

G0302-5G G0302-25G
TCG0302-5GEA 67.06 CAD
TCG0302-5G TCG0302-25G
D-Glucoronic acid ≥96.0% (by HPLC, titration analysis)
D-Glucoronic acid

CAS Number: 6556-12-3
MDL Number: MFCD00077778
Molecular Formula: C6H10O7
Molecular Weight: 194.14
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 153
Specific rotation [a]20/D: 36 deg (C=6, H2O)

Formula: C₆H₁₀O₇
Melting Pt: 153 °C
MDL Number: MFCD00166981
CAS Number: 6556-12-3

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR