(S)-(+)-Glycidyl butyrate ≥98.0%

Supplier: TCI America

Synonyms: (S)-Oxiran-2-ylmethyl butyrate

G0283-25G G0283-5G
TCG0283-25GEA 547.15 CAD
TCG0283-25G TCG0283-5G
(S)-(+)-Glycidyl butyrate ≥98.0%
(S)-(+)-Glycidyl butyrate

CAS Number: 65031-96-1
MDL Number: MFCD00273369
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 84
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: 31 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: Butyric Acid (S)-Glycidyl Ester

Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
Boiling Pt: 90 °C
Density: 1.04 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00273369
CAS Number: 65031-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d57c90f2c1908960c77526b9f86d928796603a6&vid=3d57c90f2c1908960c77526b9f86d928796603a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR