(R)-(-)-Glycidyl butyrate ≥98.0%

Supplier: TCI America

G0282-25G G0282-5G
TCG0282-25GEA 295.46 CAD
TCG0282-25G TCG0282-5G
(R)-(-)-Glycidyl butyrate ≥98.0%
(R)-(-)-Glycidyl butyrate

CAS Number: 60456-26-0
MDL Number: MFCD00075120
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 84
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: -31 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: Butyric Acid (R)-Glycidyl Ester

Formula: C₇H₁₂O₃
Boiling Pt: 90 °C
Density: 1.04 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00075120
CAS Number: 60456-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1da98236ae5d4890e216c06fddd6c0ec2cd6b879&vid=1da98236ae5d4890e216c06fddd6c0ec2cd6b879&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR