47% in toluene

Supplier: TCI America

G0264-25G G0264-500G G0264-100G
TCG0264-25GEA 79.32 CAD
TCG0264-25G TCG0264-500G TCG0264-100G
47% in toluene
Ethyl glyoxylate

CAS Number: 924-44-7
MDL Number: MFCD00044009
Molecular Formula: C4H6O3
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 110
Flash Point (°C): 7
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Ethyl Oxoacetate Polymer form, Glyoxalic Acid Ethyl Ester Polymer form, Glyoxylic Acid Ethyl Ester Polymer form

Formula: C₄H₆O₃
Boiling Pt: 110 °C
Density: 1.03 (20 °C)
Flash Pt: 7 °C
MDL Number: MFCD00044009
CAS Number: 924-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR