Glycinamide hydrochloride (H-Gly-NH2.HCl) ≥98.0%

Supplier: TCI America

Synonyms: H-Gly-NH2.HCl

G0253-500G G0253-25G G0253-100G
TCG0253-500GEA 235.42 CAD
TCG0253-500G TCG0253-025G TCG0253-100G
Glycinamide hydrochloride (H-Gly-NH2.HCl) ≥98.0%
Glycinamide hydrochloride (H-Gly-NH2.HCl)

CAS Number: 1668-10-6
MDL Number: MFCD00013008
Molecular Formula: C2H6N2O
Molecular Weight: 110.54
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₂H₇ClN₂O
MW: 110.54 g/mol
MDL Number: MFCD00013008
CAS Number: 1668-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1035544208c170e2af1b28e63173091ed85df764&vid=1035544208c170e2af1b28e63173091ed85df764&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR