Ethyl glycolate ≥98.0%

Supplier: TCI America

G0242-25G G0242-100G
TCG0242-025GEA 91.64 CAD
TCG0242-025G TCG0242-100G
Ethyl glycolate ≥98.0%
Ethyl glycolate

CAS Number: 623-50-7
MDL Number: MFCD00021970
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 62
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: Glycolic Acid Ethyl Ester, Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester

Formula: C₄H₈O₃
Boiling Pt: 160 °C
Density: 1.10 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00021970
CAS Number: 623-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8f48bc2bb65f3e11e30be7d2fa35ea359e785e3&vid=b8f48bc2bb65f3e11e30be7d2fa35ea359e785e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR