Penta-O-acetyl-ɑ-D-glucopyranose ≥97.0%

Supplier: TCI America

Synonyms: 1,2,3,4,6-Penta-O-acetyl-α-D-glycopyranose, ɑ-D-Glucose Pentaacetate, 1,2,3,4,6-Penta-O-Acetyl-A-D-Glycopyranose, α-D(+)-Glucose pentaacetate, (2R,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, Pentaacetyl-ɑ-D-glucose, α-D-Glucose pentaacetate

G0225-25G G0225-250G
TCG0225-25GEA 41.08 CAD
TCG0225-25G TCG0225-010G TCG0225-250G
Penta-O-acetyl-ɑ-D-glucopyranose ≥97.0%
Penta-O-acetyl-alpha-D-glucopyranose

CAS Number: 604-68-2
MDL Number: MFCD00064071
Molecular Formula: C16H22O11
Molecular Weight: 390.34
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 113
Flash Point (°C): 150
Specific rotation [a]20/D: 106 deg (C=1.5, MeOH)


Product Categories: Bioscience, Glucose, O-Substituted Sugars
Other Product Names / Synonyms: alpha-D-Glucose Pentaacetate, Pentaacetyl-alpha-D-glucose

Formula: C₁₆H₂₂O₁₁
MW: 390.34 g/mol
Melting Pt: 113 °C
Flash Pt: 150 °C
MDL Number: MFCD00064071
CAS Number: 604-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bbf1bfddd5415793313c6b669c035e0a0b6f271&vid=5bbf1bfddd5415793313c6b669c035e0a0b6f271&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR