Tripalmitin (glycerol tripalmitate) ≥85.0%

Supplier: TCI America

Synonyms: Propane-1,2,3-triyl tripalmitate, Tripalmitin, Glycerol tripalmitate

G0213-25G G0213-250G
TCG0213-025GEA 188.02 CAD
TCG0213-025G TCG0213-250G
Tripalmitin (glycerol tripalmitate) ≥85.0%
Tripalmitin (Glycerol tripalmitate)

CAS Number: 555-44-2
MDL Number: MFCD00008995
Molecular Formula: C51H98O6
Molecular Weight: 807.34
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Boiling point (°C): 320
Melting point (°C): 66
Flash Point (°C): 66


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Tripalmitate

Formula: C₅₁H₉₈O₆
MW: 807.34 g/mol
Boiling Pt: 320 °C
Melting Pt: 66 °C
Flash Pt: 66 °C
MDL Number: MFCD00008995
CAS Number: 555-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e4b82c4e429e24451fdef8e13873e8c26ad61e3&vid=2e4b82c4e429e24451fdef8e13873e8c26ad61e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR