Glutaconaldehydedianil hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
N-(5-Anilino-2,4-pentadienylidene)aniline hydrochloride, N-[5-(Phenylamino)-2,4-pentadienylidene]aniline monohydrochloride, N-((2E,4E)-5-(Phenylamino)penta-2,4-dien-1-ylidene)aniline hydrochloride
CAS Number: 1497-49-0
MDL Number: MFCD00012630
Molecular Formula: C17H16N2
Molecular Weight: 284.79
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 140
Other Product Names / Synonyms: N-(5-Anilino-2,4-pentadienylidene)aniline Hydrochloride
Formula:
C₁₇H₁₆N₂ Melting Pt: 140 °C |
MDL Number:
MFCD00012630 CAS Number: 1497-49-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...