Methyl glycolate ≥98.0%

Supplier: TCI America

G0199-500G G0199-25G G0199-100G
TCG0199-500GEA 759.98 CAD
TCG0199-500G TCG0199-025G TCG0199-100G
Methyl glycolate ≥98.0%
Methyl glycolate

CAS Number: 96-35-5
MDL Number: MFCD00004667
Molecular Formula: C3H6O3
Molecular Weight: 90.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 67
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: Hydroxyacetic Acid Methyl Ester, Glycolic Acid Methyl Ester, Methyl Hydroxyacetate

Formula: C₃H₆O₃
Boiling Pt: 151 °C
Density: 1.19 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00004667
CAS Number: 96-35-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c5e49d6134a48bbd42af99994500314ed1ab3d8&vid=2c5e49d6134a48bbd42af99994500314ed1ab3d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR