Glycinamide hydrochloride (H-Gly-NH2.HCl) ≥98.0%

Supplier: TCI America

Synonyms: H-Gly-NH2.HCl

G0193-100G G0193-25G
TCG0193-100GEA 301.78 CAD
TCG0193-100G TCG0193-025G
Glycinamide hydrochloride (H-Gly-NH2.HCl) ≥98.0%
Glycinamide hydrochloride (H-Gly-NH2.HCl)

CAS Number: 1668-10-6
MDL Number: MFCD00013008
Molecular Formula: C2H6N2O
Molecular Weight: 110.54
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 203

Formula: C₂H₇ClN₂O
Melting Pt: 203 °C
MDL Number: MFCD00013008
CAS Number: 1668-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7100d4eaac98606c24c072fd3877e98fb30bcdd&vid=d7100d4eaac98606c24c072fd3877e98fb30bcdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR