Glyoxal 39% ∼8.8 mol/L in water
Supplier: TCI America
G0152-25G
G0152-500G
TCG0152-025GEA
29.96
CAD
TCG0152-025G
TCG0152-500G
Glyoxal 39% ∼8.8 mol/L in water
Glyoxal
CAS Number: 107-22-2
MDL Number: MFCD00006957
Molecular Formula: C2H2O2
Molecular Weight: 58.04
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.27
Formula:
C₂H₂O₂ Density: 1.27 (20 °C) |
MDL Number:
MFCD00006957 CAS Number: 107-22-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...