Sodium glycolate

Supplier: TCI America

G0111-500G G0111-25G
TCG0111-500GEA 216.46 CAD
TCG0111-500G TCG0111-025G
Sodium glycolate
Sodium glycolate

CAS Number: 2836-32-0
MDL Number: MFCD00065358
Molecular Formula: C2H4O3
Molecular Weight: 98.03
Form: Crystal
Color: White


Other Product Names / Synonyms: Glycolic Acid Sodium Salt, Hydroxyacetic Acid Sodium Salt

Formula: C₂H₃NaO₃
MW: 98.03 g/mol
MDL Number: MFCD00065358
CAS Number: 2836-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1581651203fed564336e132e1e8029dc39cb229d&vid=1581651203fed564336e132e1e8029dc39cb229d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR