Diacetin (contains Mono-, Tri-, Glycerol)
Supplier: TCI America
G0077-25G
G0077-500G
TCG0077-025GEA
30.02
CAD
TCG0077-025G
TCG0077-500G
Diacetin (contains Mono-, Tri-, Glycerol)
Diacetin
CAS Number: 25395-31-7
MDL Number: MFCD00008717
Molecular Formula: C7H12O5
Molecular Weight: 176.17
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 261
Flash Point (°C): 146
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: Glycerol alpha,alpha'-Diacetate
Formula:
C₇H₁₂O₅ Boiling Pt: 261 °C Density: 1.18 (20 °C) Flash Pt: 146 °C |
MDL Number:
MFCD00008717 CAS Number: 25395-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...