γ-L-Glutamyl-4-nitroanilide (GPNA) hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
G0065-1G
G0065-5G
TCG0065-001GEA
225.94
CAD
TCG0065-001G
TCG0065-005G
γ-L-Glutamyl-4-nitroanilide (GPNA) hydrate ≥98.0% (by HPLC, titration analysis)
γ-L-Glutamyl-4-nitroanilide (GPNA) hydrate
CAS Number: 7300-59-6
MDL Number: MFCD00036218
Molecular Formula: C11H13N3O5
Molecular Weight: 267.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 178
Specific rotation [a]20/D: 32 deg (C=2, 0.5mol/L HCl)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: Nomega-(4-Nitrophenyl)-L-glutamine
Formula:
C₁₁H₁₃N₃O₅ Melting Pt: 178 °C |
MDL Number:
MFCD00036218 CAS Number: 7300-59-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...