D-(+)-Glucurono-6,3-lactone ≥99.0% (by titrimetric analysis)
Supplier: TCI America
G0055-25G
G0055-500G
G0055-100G
TCG0055-025GEA
44.24
CAD
TCG0055-025G
TCG0055-500G
TCG0055-100G
D-(+)-Glucurono-6,3-lactone ≥99.0% (by titrimetric analysis)
D(+)-Glucurono-6,3-lactone
CAS Number: 32449-92-6
MDL Number: MFCD00135622
Molecular Formula: C6H8O6
Molecular Weight: 176.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 178
Specific rotation [a]20/D: 18.5 deg (C=5, H2O)
Product Categories: Bioscience, Sugar Acids, Glucuronic Acids
Other Product Names / Synonyms: D-Glucuronic Acid Lactone
Formula:
C₆H₈O₆ Melting Pt: 178 °C |
MDL Number:
MFCD00135622 CAS Number: 32449-92-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...