D-(+)-Glucosamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0044-500G G0044-25G
TCG0044-500GEA 248.41 CAD
TCG0044-500G TCG0044-025G
D-(+)-Glucosamine hydrochloride ≥98.0% (by titrimetric analysis)
D-(+)-Glucosamine hydrochloride

CAS Number: 66-84-2
MDL Number: MFCD00135831
Molecular Formula: C6H13NO5
Molecular Weight: 215.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 194
Specific rotation [a]20/D: 72 deg (C=1, H2O)


Product Categories: Bioscience, Glucosamine, Aminosugars, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride

Formula: C₆H₁₃NO₅
Melting Pt: 194 °C
MDL Number: MFCD00135831
CAS Number: 66-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9127a0c70599f989b391bf09127dc564544e81a&vid=f9127a0c70599f989b391bf09127dc564544e81a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR