Geranyl acetate ≥70.0%
Supplier: TCI America
G0028-25ML
G0028-500ML
G0028-100ML
TCG0028-025MLEA
30.56
CAD
TCG0028-025ML
TCG0028-500ML
TCG0028-100ML
Geranyl acetate ≥70.0%
Geranyl acetate
CAS Number: 105-87-3
MDL Number: MFCD00015037
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: >70.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 118
Specific Gravity (20/20): 0.91
Formula:
C₁₂H₂₀O₂ Boiling Pt: 120 °C Density: 0.91 (20 °C) Flash Pt: 118 °C |
MDL Number:
MFCD00015037 CAS Number: 105-87-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...