4'-Fluoro-2'-hydroxyacetophenone ≥97.0%
Supplier: TCI America
F0723-5G
F0723-25G
TCF0723-5GEA
97.96
CAD
TCF0723-5G
TCF0723-25G
4'-Fluoro-2'-hydroxyacetophenone ≥97.0%
4'-Fluoro-2'-hydroxyacetophenone
CAS Number: 1481-27-2
MDL Number: MFCD00143203
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 99
Freezing point (°C): 27
Formula:
C₈H₇FO₂ Flash Pt: 99 °C |
MDL Number:
MFCD00143203 CAS Number: 1481-27-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...