5-Fluoroindole-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
F0716-1G
F0716-5G
TCF0716-1GEA
91.64
CAD
TCF0716-1G
TCF0716-5G
5-Fluoroindole-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Fluoroindole-2-carboxylic acid
CAS Number: 399-76-8
MDL Number: MFCD00005612
Molecular Formula: C9H6FNO2
Molecular Weight: 179.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 249
Formula:
C₉H₆FNO₂ Melting Pt: 249 °C |
MDL Number:
MFCD00005612 CAS Number: 399-76-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...