3-Fluoro-4-methylphenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)

Supplier: TCI America

F0712-1G F0712-5G
TCF0712-1GEA 52.14 CAD
TCF0712-1G TCF0712-5G
3-Fluoro-4-methylphenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
3-Fluoro-4-methylphenylboronic acid

CAS Number: 168267-99-0
MDL Number: MFCD02683115
Molecular Formula: C7H8BFO2
Molecular Weight: 153.95
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 237


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 3-Fluoro-4-methylbenzeneboronic Acid, 3-Fluoro-p-tolylboronic Acid

Formula: C₇H₈BFO₂
Melting Pt: 237 °C
MDL Number: MFCD02683115
CAS Number: 168267-99-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9bd5252e6a5eb5b0f89c0c09797872788318b24&vid=d9bd5252e6a5eb5b0f89c0c09797872788318b24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR