4-Fluoro-3-methylphenylboronic acid ≥97% (contains varying amounts of Anhydride)
Supplier: TCI America
F0697-25G
F0697-5G
TCF0697-25GEA
429.76
CAD
TCF0697-25G
TCF0697-5G
4-Fluoro-3-methylphenylboronic acid ≥97% (contains varying amounts of Anhydride)
4-Fluoro-3-methylphenylboronic acid
CAS Number: 139911-27-6
MDL Number: MFCD01863527
Molecular Formula: C7H8BFO2
Molecular Weight: 153.95
Form: Crystal
Color: White
Melting point (°C): 217
Other Product Names / Synonyms: 4-Fluoro-3-methylbenzeneboronic Acid, 4-Fluoro-m-tolylboronic Acid
Formula:
C₇H₈BFO₂ Melting Pt: 217 °C |
MDL Number:
MFCD01863527 CAS Number: 139911-27-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...