5-Fluoro-2-nitrobenzoic acid ≥98.0%

Supplier: TCI America

F0694-25G F0694-5G
TCF0694-25GEA 266.07 CAD
TCF0694-25G TCF0694-5G
5-Fluoro-2-nitrobenzoic acid ≥98.0%
5-Fluoro-2-nitrobenzoic acid

CAS Number: 320-98-9
MDL Number: MFCD00055635
Molecular Formula: C7H4FNO4
Molecular Weight: 185.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 136

Formula: C₇H₄FNO₄
MW: 185.11
Melting Pt: 136 °C
MDL Number: MFCD00055635
CAS Number: 320-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bf36792394f34ca5d9c25aed3fd2a153e3395c6&vid=7bf36792394f34ca5d9c25aed3fd2a153e3395c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR