4-Fluoro-2-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0680-5G F0680-1G
TCF0680-5GEA 137.46 CAD
TCF0680-5G TCF0680-1G
4-Fluoro-2-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Fluoro-2-methoxybenzoic acid

CAS Number: 395-82-4
MDL Number: MFCD00673010
Molecular Formula: C8H7FO3
Molecular Weight: 170.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 137


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-o-anisic Acid

Formula: C₈H₇FO₃
Melting Pt: 137 °C
MDL Number: MFCD00673010
CAS Number: 395-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609f1c46fbe8da300ba7bcfa8e956b695f65643a&vid=609f1c46fbe8da300ba7bcfa8e956b695f65643a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR