1-(4-Formylphenyl)pyrrolidine ≥98.0%

Supplier: TCI America

Synonyms: 4-(1-Pyrrolidino)benzaldehyde, 4-(1-Pyrrolidinyl)benzaldehyde

F0679-1G F0679-5G
TCF0679-1GEA 63.2 CAD
TCF0679-1G TCF0679-5G
1-(4-Formylphenyl)pyrrolidine ≥98.0%
1-(4-Formylphenyl)pyrrolidine

CAS Number: 51980-54-2
MDL Number: MFCD00015469
Molecular Formula: C11H13NO
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: 4-(1-Pyrrolidinyl)benzaldehyde

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00015469
CAS Number: 51980-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99be7b76218cd7d0db2e6806e7fecccd1dc97178&vid=99be7b76218cd7d0db2e6806e7fecccd1dc97178&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR