3-Fluoro-4-nitrobenzoic acid ≥98.0%
Supplier: TCI America
F0670-5G
F0670-25G
TCF0670-5GEA
55.3
CAD
TCF0670-5G
TCF0670-25G
3-Fluoro-4-nitrobenzoic acid ≥98.0%
3-Fluoro-4-nitrobenzoic acid
CAS Number: 403-21-4
MDL Number: MFCD01862092
Molecular Formula: C7H4FNO4
Molecular Weight: 185.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 175
Formula:
C₇H₄FNO₄ MW: 185.11 Melting Pt: 175 °C |
MDL Number:
MFCD01862092 CAS Number: 403-21-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...