6-Fluoro-1-indanone ≥98.0%

Supplier: TCI America

F0668-5G F0668-1G
TCF0668-5GEA 395 CAD
TCF0668-5G TCF0668-1G
6-Fluoro-1-indanone ≥98.0%
6-Fluoro-1-indanone

CAS Number: 1481-32-9
MDL Number: MFCD01318147
Molecular Formula: C9H7FO
Molecular Weight: 150.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 60
Melting point (°C): 60

Formula: C₉H₇FO
Boiling Pt: 60 °C
Melting Pt: 60 °C
MDL Number: MFCD01318147
CAS Number: 1481-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d08324bd7a2b0c8f6876e3b96815b5f19572f7a9&vid=d08324bd7a2b0c8f6876e3b96815b5f19572f7a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR