4-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid

F0666-5G F0666-1G
TCF0666-5GEA 395 CAD
TCF0666-5G TCF0666-1G
4-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-2-methylphenylboronic acid

CAS Number: 139911-29-8
MDL Number: MFCD02093072
Molecular Formula: C7H8BFO2
Molecular Weight: 153.95
Form: Crystal
Color: White
Melting point (°C): 201


Other Product Names / Synonyms: 4-Fluoro-2-methylbenzeneboronic Acid

Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 201 °C
MDL Number: MFCD02093072
CAS Number: 139911-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5edc9e15a31d7f161e26115bc02f862873741b6&vid=f5edc9e15a31d7f161e26115bc02f862873741b6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR