2'-Fluoro-4'-methoxyacetophenone ≥98.0%

Supplier: TCI America

F0662-1G F0662-5G
TCF0662-1GEA 60.04 CAD
TCF0662-1G TCF0662-5G
2'-Fluoro-4'-methoxyacetophenone ≥98.0%
2'-Fluoro-4'-methoxyacetophenone

CAS Number: 74457-86-6
MDL Number: MFCD00042290
Molecular Formula: C9H9FO2
Molecular Weight: 168.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 85
Melting point (°C): 52
Flash Point (°C): 113

Formula: C₉H₉FO₂
Boiling Pt: 85 °C
Melting Pt: 52 °C
Flash Pt: 113 °C
MDL Number: MFCD00042290
CAS Number: 74457-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526e51ccbbd0e9f1f999daa2e22ba02df40626b6&vid=526e51ccbbd0e9f1f999daa2e22ba02df40626b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR