N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0652-5G F0652-25G
TCF0652-5GEA 66.36 CAD
TCF0652-5G TCF0652-25G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine

CAS Number: 103213-32-7
MDL Number: MFCD00038538
Molecular Formula: C37H31NO4S
Molecular Weight: 585.72
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 175
Specific rotation [a]20/D: 18 deg (C=1, THF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-S-trityl-L-cysteine, Fmoc-Cys(Trt)-OH

Formula: C₃₇H₃₁NO₄S
Melting Pt: 175 °C
MDL Number: MFCD00038538
CAS Number: 103213-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bb74954bcf714f8c9f67bdf022afe3ba34f1250&vid=4bb74954bcf714f8c9f67bdf022afe3ba34f1250&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR