4-Fluorophenyl methyl sulfone ≥98.0%

Supplier: TCI America

F0651-5G F0651-25G
TCF0651-5GEA 115.34 CAD
TCF0651-5G TCF0651-25G
4-Fluorophenyl methyl sulfone ≥98.0%
4-Fluorophenyl methyl sulfone

CAS Number: 455-15-2
MDL Number: MFCD00039753
Molecular Formula: C7H7FO2S
Molecular Weight: 174.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80

Formula: C₇H₇FO₂S
Melting Pt: 80 °C
MDL Number: MFCD00039753
CAS Number: 455-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb7a573f176f8b0675a59033ce9bc23af7a8352c&vid=fb7a573f176f8b0675a59033ce9bc23af7a8352c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR