3-Fluoro-p-anisidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0648-5G F0648-25G
TCF0648-5GEA 82.16 CAD
TCF0648-5G TCF0648-25G
3-Fluoro-p-anisidine ≥98.0% (by GC, titration analysis)
3-Fluoro-p-anisidine

CAS Number: 366-99-4
MDL Number: MFCD00075040
Molecular Formula: C7H8FNO
Molecular Weight: 141.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 82
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Amino-2-fluoroanisole, 3-Fluoro-p-anisidine, 4-Amino-2-fluoro-1-methoxybenzene

Formula: C₇H₈FNO
Boiling Pt: 135 °C
Melting Pt: 82 °C
Flash Pt: 110 °C
MDL Number: MFCD00075040
CAS Number: 366-99-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95cdf8fb9bbe4aa2ec3580911b0c8c2166477a02&vid=95cdf8fb9bbe4aa2ec3580911b0c8c2166477a02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR