5-Fluoro-2-nitrobenzaldehyde ≥98.0%

Supplier: TCI America

F0645-5G F0645-25G
TCF0645-5GEA 74.26 CAD
TCF0645-5G TCF0645-25G
5-Fluoro-2-nitrobenzaldehyde ≥98.0%
5-Fluoro-2-nitrobenzaldehyde

CAS Number: 395-81-3
MDL Number: MFCD00153175
Molecular Formula: C7H4FNO3
Molecular Weight: 169.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 153
Melting point (°C): 94

Formula: C₇H₄FNO₃
Boiling Pt: 153 °C
Melting Pt: 94 °C
MDL Number: MFCD00153175
CAS Number: 395-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e8828c79077466fbebb5c9b63f020fe736e4a39&vid=da5267597344e2e18cf73a175e9db080cf442788&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR