5-Fluoro-o-cresol ≥98.0%

Supplier: TCI America

F0632-1G F0632-5G
TCF0632-1GEA 85.95 CAD
TCF0632-1G TCF0632-5G
5-Fluoro-o-cresol ≥98.0%
5-Fluoro-o-cresol

CAS Number: 452-85-7
MDL Number: MFCD00673009
Molecular Formula: C7H7FO
Molecular Weight: 126.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 66
Specific Gravity (20/20): 1.17


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-2-hydroxytoluene, 5-Fluoro-2-methylphenol

Formula: C₇H₇FO
Boiling Pt: 66 °C
Density: 1.17 (20 °C)
MDL Number: MFCD00673009
CAS Number: 452-85-7
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94f6f104ab201a1567e4d2a68dc277e594674ea1&vid=94f6f104ab201a1567e4d2a68dc277e594674ea1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR